siRNA Details
Gene Symbol: Neurl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163480.1 | NP_001156952.1 | 3 | 728 | ||
NM_021360.4 | NP_067335.4 | 3 | 1206 | |||
GenBank mRNA | AF400063.1 | - | 3 | 777 | ||
AF401228.1 | - | 3 | 663 | |||
AK010787.1 | - | 3 | 875 | |||
AK046382.1 | - | 3 | 1005 | |||
AK078529.1 | - | 1 | 263 | |||
BC058386.1 | - | 3 | 1031 | 6831952 | ||
BC099702.1 | - | 3 | 1206 | 6825751 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | Y15160.1 | - | 559 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
