siRNA Details
Gene Symbol: Neurod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010894.2 | NP_035024.1 | 2 | 487 | ||
GenBank mRNA | AK005073.1 | - | 2 | 488 | ||
AK018781.1 | - | 2 | 486 | |||
AK028055.1 | - | 2 | 491 | |||
BC018241.1 | - | 2 | 463 | 4511370 | ||
BC094611.1 | - | 2 | 427 | 30361598 | ||
U28068.1 | - | 1 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
