siRNA Details
Gene Symbol: Neurod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010895.3 | NP_035025.3 | 2 | 1292 | ||
GenBank mRNA | AK158767.1 | - | 2 | 1424 | ||
BC058965.1 | - | 2 | 1241 | 6817440 | ||
D83507.1 | - | 2 | 1108 | |||
U58471.1 | - | 2 | 1304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
