siRNA Details
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010901.2 | NP_035031.2 | 6 | 2153 | ||
GenBank mRNA | AK084848.1 | - | 1 | 395 | ||
AK131919.1 | - | 4 | 565 | |||
AK134054.1 | - | 6 | 2072 | |||
AK147708.1 | - | 6 | 2123 | |||
AK220187.1 | - | 6 | 1925 | |||
BC021835.1 | - | 3 | 357 | |||
BC120900.1 | - | 6 | 1872 | 40085687 | ||
BC120901.1 | - | 6 | 1872 | 40085688 | ||
D85612.1 | - | 6 | 1869 | |||
EU887626.1 | - | 6 | 1841 | |||
EU887627.1 | - | 6 | 1841 | |||
EU887629.1 | - | 6 | 1865 | |||
EU887630.1 | - | 6 | 1865 | |||
U28807.1 | - | 6 | 1832 |

More Information
Other Species Targeted

Gene Ontology Categories:
