siRNA Details
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010901.2 | NP_035031.2 | 5 | 1928 | ||
GenBank mRNA | AK131919.1 | - | 3 | 340 | ||
AK134054.1 | - | 5 | 1847 | |||
AK147708.1 | - | 5 | 1898 | |||
AK220187.1 | - | 5 | 1700 | |||
BC021835.1 | - | 2 | 132 | |||
BC120900.1 | - | 5 | 1647 | 40085687 | ||
BC120901.1 | - | 5 | 1647 | 40085688 | ||
D85612.1 | - | 5 | 1644 | |||
EU887626.1 | - | 5 | 1616 | |||
EU887627.1 | - | 5 | 1616 | |||
EU887629.1 | - | 5 | 1640 | |||
EU887630.1 | - | 5 | 1640 | |||
U28807.1 | - | 5 | 1607 |
