siRNA Details
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010901.2 | NP_035031.2 | 9 | 3140 | ||
GenBank mRNA | AK131919.1 | - | 7 | 1552 | ||
AK134054.1 | - | 9 | 3059 | |||
AK161185.1 | - | 1 | 190 | |||
AK220187.1 | - | 9 | 2913 | |||
BC021835.1 | - | 6 | 1344 | |||
BC120900.1 | - | 9 | 2859 | 40085687 | ||
BC120901.1 | - | 9 | 2859 | 40085688 | ||
D85612.1 | - | 9 | 2856 | |||
EU887626.1 | - | 9 | 2828 | |||
EU887627.1 | - | 9 | 2828 | |||
EU887629.1 | - | 9 | 2852 | |||
EU887630.1 | - | 9 | 2852 | |||
U28807.1 | - | 9 | 2819 |
