siRNA Details
Gene Symbol: Nfe2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130450.1 | NP_001123922.1 | 3 | 1216 | ||
NM_001130451.1 | NP_001123923.1 | 2 | 150 | |||
NM_001130452.1 | NP_001123924.1 | 2 | 117 | |||
NM_001130453.1 | NP_001123925.1 | 2 | 167 | |||
NM_001130454.1 | NP_001123926.1 | 2 | 134 | |||
NM_008686.3 | NP_032712.2 | 3 | 1274 | |||
GenBank mRNA | AF071084.1 | - | 2 | 620 | ||
AK131183.1 | - | 3 | 1209 | |||
AK148471.1 | - | 2 | 134 | |||
AK152040.1 | - | 3 | 1255 | |||
AK153529.1 | - | 3 | 1256 | |||
AK154167.1 | - | 3 | 1185 | |||
AK184212.1 | - | 2 | 284 | |||
AK194563.1 | - | 1 | 96 | |||
AK201732.1 | - | 2 | 275 | |||
BC043063.1 | - | 2 | 856 | 5705669 | ||
BC047283.1 | - | 2 | 616 | 6489220 | ||
BC099895.1 | - | 3 | 1240 | |||
CF163519.1 | - | 2 | 162 | |||
CJ144781.1 | - | 2 | 123 | |||
CJ152359.1 | - | 2 | 138 | |||
X78709.1 | - | 2 | 652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
