siRNA Details
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008691.2 | NP_032717.2 | 3 | 1939 | ||
GenBank mRNA | AK051296.1 | - | 3 | 1782 | ||
AK051696.1 | - | 3 | 1779 | |||
AK164318.1 | - | 3 | 1782 | |||
BC058333.1 | - | 3 | 1772 | |||
BC119602.2 | - | 3 | 1747 | |||
BC128564.1 | - | 3 | 1742 | |||
DQ201636.1 | - | 3 | 1757 |
