siRNA Details
Gene Symbol: Nfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010911.2 | NP_035041.2 | 6 | 663 | ||
GenBank mRNA | AJ222660.1 | - | 6 | 662 | ||
AK005724.1 | - | 6 | 652 | |||
AK075575.1 | - | 6 | 661 | |||
AK132838.1 | - | 6 | 664 | |||
AK159828.1 | - | 6 | 652 | |||
AK183689.1 | - | 2 | 69 | |||
AK206231.1 | - | 2 | 69 | |||
AK206381.1 | - | 2 | 69 | |||
AK206572.1 | - | 2 | 69 | |||
AK207148.1 | - | 2 | 69 | |||
AK207847.1 | - | 2 | 69 | |||
AK207895.1 | - | 2 | 69 | |||
AK211411.1 | - | 2 | 69 | |||
AK211628.1 | - | 2 | 69 | |||
AK212905.1 | - | 2 | 69 | |||
AK216163.1 | - | 1 | 69 | |||
AK220082.1 | - | 1 | 69 | |||
BC010586.1 | - | 6 | 641 | 3711352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
