siRNA Details
Gene Symbol: Nfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010911.2 | NP_035041.2 | 4 | 449 | ||
GenBank mRNA | AJ222660.1 | - | 4 | 448 | ||
AK005724.1 | - | 4 | 438 | |||
AK075575.1 | - | 4 | 447 | |||
AK084831.1 | - | 4 | 418 | |||
AK132838.1 | - | 4 | 450 | |||
AK148527.1 | - | 4 | 450 | |||
AK159828.1 | - | 4 | 438 | |||
BC010586.1 | - | 4 | 427 | 3711352 | ||
BC012651.1 | - | 4 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
