siRNA Details
Gene Symbol: Ngp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008694.2 | NP_032720.2 | 3, 4 | 419 | ||
GenBank mRNA | AK009649.1 | - | 3, 4 | 373 | ||
AK165210.1 | - | 3, 4 | 372 | |||
BC119403.1 | - | 3, 4 | 390 | 8734152 | ||
BC119405.1 | - | 3, 4 | 390 | 8734154 | ||
L37297.1 | - | 3, 4 | 424 | |||
U95002.1 | - | 3, 4 | 378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
