siRNA Details
Gene Symbol: Nin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081453.1 | NP_001074922.1 | 21 | 5107 | ||
NM_001286079.2 | NP_001273008.1 | 22 | 5289 | |||
NM_001286080.2 | NP_001273009.1 | 23 | 5336 | |||
NM_008697.4 | NP_032723.2 | 23 | 5336 | |||
GenBank mRNA | AK129394.1 | - | 23 | 5226 | ||
AK134860.1 | - | 21 | 3180 | |||
AK147187.1 | - | 6 | 2092 | |||
AK162150.1 | - | 22 | 3160 | |||
AK167264.1 | - | 19 | 4202 | |||
AY515727.1 | - | 21 | 5086 | |||
BC137789.1 | - | 22 | 5174 | 8860808 | ||
BC144913.1 | - | 23 | 5221 | 9053461 | ||
BC144914.1 | - | 23 | 5221 |
