siRNA Details
Gene Symbol: Nkx2-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010920.2 | NP_035050.2 | 2 | 1074 | ||
GenBank mRNA | AF030113.1 | - | 2 | 885 | ||
AK006401.1 | - | 2 | 930 | |||
AK007304.1 | - | 2 | 857 | |||
BC100425.1 | - | 2 | 892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
