siRNA Details
Gene Symbol: Mycn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008709.3 | NP_032735.3 | 3 | 1389 | ||
GenBank mRNA | AK144126.1 | - | 3 | 1390 | ||
AK146696.1 | - | 3 | 1390 | |||
AK146913.1 | - | 3 | 1387 | |||
BC005453.1 | - | 3 | 1273 | 3495446 | ||
BC049783.1 | - | 2 | 1318 | 6517457 | ||
M36277.1 | - | 1165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
