siRNA Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291128.1 | NP_001278057.1 | 3 | 1075 | ||
NM_001291129.1 | NP_001278058.1 | 3 | 1063 | |||
NM_001291130.1 | NP_001278059.1 | 3 | 1014 | |||
NM_010923.3 | NP_035053.1 | 3 | 1068 | |||
NM_180960.3 | NP_851291.1 | 2 | 987 | |||
GenBank mRNA | AB004048.1 | - | 3 | 1275 | ||
AK003004.1 | - | 3 | 1032 | |||
AK160626.1 | - | 3 | 1036 | |||
BC036984.1 | - | 3 | 1026 | 4981517 | ||
BU705167.1 | - | 1 | 329 | |||
CJ302750.1 | - | 1 | 242 | |||
GQ184462.1 | - | 3 | 970 | |||
X83568.1 | - | 3 | 836 | |||
X83569.1 | - | 2 | 943 | |||
X83570.1 | - | 3 | 1024 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
