siRNA Details
Gene Symbol: Ints6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008715.2 | NP_032741.2 | 5 | 995 | ||
GenBank mRNA | AK149711.1 | - | 5 | 1023 | ||
AK172178.1 | - | 5 | 1023 | |||
AK186108.1 | - | 3 | 327 | |||
BC059263.1 | - | 5 | 995 | 6409580 |
