siRNA Details
Gene Symbol: Nov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010930.4 | NP_035060.1 | 4 | 794 | ||
GenBank mRNA | AK039481.1 | - | 4 | 785 | ||
AK081310.1 | - | 4 | 812 | |||
AK081944.1 | - | 4 | 1020 | |||
BC003774.1 | - | 4 | 800 | 3494896 | ||
X96585.1 | - | 4 | 794 | |||
Y09257.1 | - | 4 | 782 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
