siRNA Details
Gene Symbol: Npm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252260.1 | NP_001239189.1 | 8 | 824 | ||
NM_001252261.1 | NP_001239190.1 | 8 | 863 | |||
NM_008722.3 | NP_032748.1 | 8 | 863 | |||
GenBank mRNA | AK005420.1 | - | 8 | 734 | ||
AK005498.1 | - | 8 | 733 | |||
AK028253.1 | - | 8 | 734 | |||
AK028370.1 | - | 8 | 733 | |||
AK088923.1 | - | 8 | 733 | |||
AK135517.1 | - | 8 | 734 | |||
AK145124.1 | - | 8 | 733 | |||
AK145813.1 | - | 8 | 734 | |||
AK153901.1 | - | 8 | 734 | |||
AK165860.1 | - | 8 | 735 | |||
BC054755.1 | - | 8 | 716 | 6411700 | ||
BC085278.1 | - | 8 | 694 | 30438901 | ||
BC089546.1 | - | 8 | 733 | 30505980 | ||
BC090843.1 | - | 8 | 716 | 6820557 | ||
BC092378.1 | - | 8 | 698 | 5710278 | ||
BU513690.1 | - | 8 | 681 | |||
CX567338.1 | - | 3 | 148 | |||
GU214027.1 | - | 8 | 737 | |||
M33212.1 | - | 8 | 714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
