siRNA Details
Gene Symbol: Nppc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010933.5 | NP_035063.1 | 3 | 803 | ||
GenBank mRNA | AK020093.1 | - | 3 | 809 | ||
AK036176.1 | - | 3 | 807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
