siRNA Details
Gene Symbol: Ctnnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008729.2 | NP_032755.2 | 23 | 4079 | ||
GenBank mRNA | AK140454.1 | - | 13 | 1697 | ||
AK140675.1 | - | 1 | 1083 | |||
AK140960.1 | - | 8 | 954 | |||
AK164175.1 | - | 4 | 552 | |||
AK164304.1 | - | 6 | 630 | |||
BC138260.1 | - | 23 | 3624 | 8861279 | ||
BC138261.1 | - | 23 | 3624 | 8861280 | ||
BC145217.1 | - | 22 | 3546 | 9053768 | ||
U90331.1 | - | 23 | 4082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
