siRNA Details
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164226.1 | NP_001157698.1 | 4, 5 | 630 | ||
NM_001164227.1 | NP_001157699.1 | 4, 5 | 630 | |||
NM_001164228.1 | NP_001157700.1 | 4, 5 | 627 | |||
NM_001164229.1 | NP_001157701.1 | 5, 6 | 704 | |||
NM_001164230.1 | NP_001157702.1 | 4, 5 | 627 | |||
NM_010938.4 | NP_035068.3 | 4, 5 | 627 | |||
GenBank mRNA | AF098077.1 | - | 4, 5 | 562 | ||
AK014494.1 | - | 4, 5 | 591 | |||
AK029034.1 | - | 4, 5 | 608 | |||
AK037697.1 | - | 4, 5 | 588 | |||
AK050861.1 | - | 4, 5 | 544 | |||
AK082580.1 | - | 4, 5 | 628 | |||
AK134483.1 | - | 4, 5 | 544 | |||
AK135673.1 | - | 5, 6 | 707 | |||
BC005410.1 | - | 3, 4 | 448 | 3496661 |
