siRNA Details
Gene Symbol: Nrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190187.1 | NP_001177116.1 | 10 | 1953 | ||
NM_001190188.1 | NP_001177117.1 | 9 | 1908 | |||
NM_008734.3 | NP_032760.1 | 10 | 1956 | |||
GenBank mRNA | AF010130.1 | - | 10 | 1956 | ||
AK014399.1 | - | 3 | 177 | |||
BC137838.1 | - | 10 | 1693 | 8860857 | ||
BC144964.1 | - | 9 | 1651 | 9053512 |

More Information
Other Species Targeted

Gene Ontology Categories:
