siRNA Details
Gene Symbol: Nrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077403.1 | NP_001070871.1 | 12 | 2822 | ||
NM_001077404.1 | NP_001070872.1 | 12 | 2822 | |||
NM_001077405.1 | NP_001070873.1 | 12 | 2822 | |||
NM_001077406.1 | NP_001070874.1 | 12 | 2822 | |||
NM_001077407.1 | NP_001070875.1 | 12 | 2822 | |||
NM_010939.2 | NP_035069.2 | 12 | 2822 | |||
GenBank mRNA | AF483506.1 | - | 12 | 1934 | ||
AF483507.1 | - | 12 | 1934 | |||
AK141234.1 | - | 2 | 179 | |||
AK149643.1 | - | 12 | 2749 | |||
AK153553.1 | - | 12 | 2211 | |||
AK190084.1 | - | 3 | 232 | |||
AK196919.1 | - | 3 | 222 | |||
BC029876.1 | - | 4 | 590 | |||
BC057028.1 | - | 12 | 1882 | |||
BC098200.1 | - | 12 | 1935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
