siRNA Details
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198587.3 | NP_001185516.2 | 6 | 1922 | ||
NM_172544.3 | NP_766132.2 | 5 | 812 | |||
GenBank mRNA | AK032126.1 | - | 5 | 804 | ||
AK078216.1 | - | 7 | 2234 | |||
AK082055.1 | - | 7 | 2234 | |||
AK129205.1 | - | 4 | 685 | |||
BC060719.1 | - | 7 | 1497 | 6406001 | ||
CF744587.1 | - | 3 | 429 |
