siRNA Details
Gene Symbol: Ntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010947.3 | NP_035077.1 | 1 | 706 | ||
GenBank mRNA | AF128866.1 | - | 1 | 546 | ||
AF149094.1 | - | 1 | 342 | |||
AF152418.1 | - | 1 | 461 | |||
AK031199.1 | - | 1 | 851 | |||
AK083904.1 | - | 1 | 725 | |||
AK141856.1 | - | 1 | 617 | |||
BC053038.1 | - | 1 | 829 | |||
BC094362.1 | - | 1 | 706 | 6830466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
