siRNA Details
Gene Symbol: Ntsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018766.2 | NP_061236.1 | 3 | 1445 | ||
GenBank mRNA | AB017027.1 | - | 3 | 1446 | ||
BC087730.1 | - | 3 | 1370 | |||
BC138630.1 | - | 3 | 1194 | 8861650 | ||
BC138633.1 | - | 3 | 1194 | 8861653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
