siRNA Details
Gene Symbol: Dusp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008748.3 | NP_032774.1 | 5 | 1011 | ||
GenBank mRNA | BC052705.1 | - | 5 | 769 | 5697787 | |
X95518.1 | - | 4 | 654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
