siRNA Details
Gene Symbol: Oaz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301307.1 | NP_001288236.1 | 2 | 362 | ||
NM_010952.3 | NP_035082.1 | 2 | 365 | |||
GenBank mRNA | AF057298.1 | - | 2 | 318 | ||
AK146074.1 | - | 2 | 335 | |||
AK160444.1 | - | 2 | 335 | |||
AK168662.1 | - | 2 | 335 | |||
AK168853.1 | - | 2 | 335 | |||
AK169243.1 | - | 2 | 365 | |||
AK181089.1 | - | 119 | ||||
BC005477.1 | - | 2 | 320 | |||
BC055442.1 | - | 2 | 345 | |||
BC083114.1 | - | 2 | 323 | |||
BC110690.1 | - | 2 | 320 | |||
CJ072367.1 | - | 2 | 332 | |||
D78643.1 | - | 1 | 19 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001301297, NM_001109899, CB586487, FQ214772, FQ211793.
Gene Ontology Categories:
