siRNA Details
Gene Symbol: Oit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010959.2 | NP_035089.1 | 2 | 174 | ||
GenBank mRNA | AF356506.1 | - | 2 | 121 | ||
AK040570.1 | - | 2 | 175 | |||
AK050081.1 | - | 2 | 140 | |||
AY180915.1 | - | 2 | 98 |
