siRNA Details
Gene Symbol: Olfr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020598.2 | NP_065623.2 | 1 | 408 | ||
GenBank mRNA | BC139721.1 | - | 1 | 408 | 40042633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
