siRNA Details
Gene Symbol: Olfr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146563.1 | NP_666774.1 | 2 | 677 | ||
GenBank mRNA | BC120866.1 | - | 2 | 576 | 40129789 | |
BC120868.1 | - | 2 | 576 | 40129791 | ||
BC144957.1 | - | 3 | 587 | |||
X89686.1 | - | 1 | 108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
