siRNA Details
Gene Symbol: Olfr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146335.1 | NP_666447.1 | 1 | 569 | ||
GenBank mRNA | BC132306.1 | - | 1 | 607 | 40130583 | |
X89683.1 | - | 1 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
