siRNA Details
Gene Symbol: Olfr25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146870.2 | NP_667081.2 | 1 | 251 | ||
GenBank mRNA | BC125306.1 | - | 1 | 251 | 40129821 | |
BC125308.1 | - | 1 | 251 | 40129823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
