siRNA Details
Gene Symbol: Olfr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146783.2 | NP_666994.2 | 1 | 516 | ||
GenBank mRNA | BC130238.1 | - | 1 | 516 | 40142117 | |
X89681.1 | - | 1 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
