siRNA Details
Gene Symbol: Olfr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147073.1 | NP_667284.1 | 1 | 310 | ||
GenBank mRNA | BC104110.1 | - | 1 | 310 | 40043836 | |
BC104111.1 | - | 1 | 310 | 40043837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
