siRNA Details
Gene Symbol: Olfr44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146830.2 | NP_667041.2 | 2 | 126 | ||
GenBank mRNA | CB173934.1 | - | 1 | 122 | ||
CB174038.1 | - | 2 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
