siRNA Details
Gene Symbol: Olfr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146934.2 | NP_667145.1 | 3 | 1077 | ||
GenBank mRNA | AF102520.1 | - | 1 | 631 | ||
AF102526.1 | - | 1 | 631 | |||
BC108947.1 | - | 1 | 842 | 40043808 |
Gene Symbol: Olfr537-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC108946.1 | - | 1 | 843 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
