siRNA Details
Gene Symbol: Olfr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146914.2 | NP_667125.1 | 2 | 576 | ||
GenBank mRNA | BC104074.1 | - | 1 | 461 | ||
BC104075.1 | - | 1 | 471 | 40041745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
