siRNA Details
Gene Symbol: Olfr58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011001.2 | NP_035131.1 | 2 | 868 | ||
GenBank mRNA | AF102536.1 | - | 1 | 424 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011001.2 | NP_035131.1 | 2 | 868 | ||
GenBank mRNA | AF102536.1 | - | 1 | 424 |