siRNA Details
Gene Symbol: Orm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008768.1 | NP_032794.1 | 2 | 210 | ||
GenBank mRNA | BC012725.1 | - | 2 | 217 | 4162961 | |
CT010399.1 | - | 2 | 178 | |||
M27008.1 | - | 2 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
