siRNA Details
Gene Symbol: Ovol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019935.3 | NP_064319.1 | 2 | 572 | ||
GenBank mRNA | AF134804.1 | - | 2 | 588 | ||
AF134805.1 | - | 2 | 554 | |||
AK003801.1 | - | 2 | 607 | |||
AK028577.1 | - | 2 | 573 | |||
AK132478.1 | - | 2 | 573 | |||
AK132505.1 | - | 2 | 611 | |||
AK132506.1 | - | 2 | 607 | |||
AK144952.1 | - | 2 | 607 | |||
BC021411.1 | - | 2 | 652 | 5006712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
