siRNA Details
Gene Symbol: P2ry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005013.2 | - | 3 | 1378 | ||
AK017378.1 | - | 3 | 1385 | |||
AK155521.1 | - | 2 | 1230 | |||
AK155562.1 | - | 2 | 1230 | |||
AK156185.1 | - | 3 | 1182 | |||
BC006613.1 | - | 3 | 1372 | 3967332 | ||
L14751.1 | - | 2 | 1171 | |||
S83099.1 | - | 1 | 902 |
