siRNA Details
Gene Symbol: Reg3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011036.1 | NP_035166.1 | 5 | 444 | ||
GenBank mRNA | BC027525.1 | - | 4 | 427 | 3471932 | |
D13509.1 | - | 417 | ||||
D63359.1 | - | 5 | 444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
