siRNA Details
Gene Symbol: Pax4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159925.1 | NP_001153397.1 | 3 | 385 | ||
NM_001159926.1 | NP_001153398.1 | 3 | 385 | |||
NM_011038.2 | NP_035168.1 | 3 | 385 | |||
GenBank mRNA | AB008912.1 | - | 3 | 360 | ||
AB010557.1 | - | 3 | 328 | |||
AB010558.1 | - | 3 | 343 | |||
AF031150.1 | - | 3 | 418 | |||
AF104231.1 | - | 2 | 140 | |||
AF145233.1 | - | 3 | 332 | |||
BC131906.1 | - | 3 | 296 | 40130183 | ||
BC131908.1 | - | 3 | 294 | 40130185 | ||
BC145168.1 | - | 3 | 296 | 9053719 | ||
BC145169.1 | - | 3 | 296 | 9053720 | ||
Y09584.1 | - | 1 | 24 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
