siRNA Details
Gene Symbol: Pcm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023662.3 | NP_076151.2 | 35 | 6040 | ||
GenBank mRNA | AB029291.1 | - | 35 | 6031 | ||
AF352180.1 | - | 34 | 5814 | |||
AF369839.1 | - | 1 | 55 | |||
AK017964.1 | - | 8 | 1159 | |||
AK020493.1 | - | 4 | 485 | |||
AK052710.1 | - | 14 | 1909 | |||
BC139033.1 | - | 35 | 5931 | 8862055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
