siRNA Details
Gene Symbol: Pcm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023662.3 | NP_076151.2 | 24 | 4369 | ||
GenBank mRNA | AB029291.1 | - | 24 | 4360 | ||
AF352180.1 | - | 23 | 4143 | |||
AK052710.1 | - | 3 | 238 | |||
BC139033.1 | - | 24 | 4260 | 8862055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
