siRNA Details
Gene Symbol: Pcnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282992.1 | NP_001269921.1 | 22 | 4257 | ||
NM_008787.3 | NP_032813.3 | 22 | 4257 | |||
GenBank mRNA | AB207233.1 | - | 22 | 4131 | ||
AB207234.1 | - | 10 | 1552 | |||
AK122275.2 | - | 7 | 1004 | |||
BC100380.1 | - | 22 | 4052 |
