siRNA Details
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291184.1 | NP_001278113.1 | 8 | 1102 | ||
NM_011048.1 | NP_035178.1 | 8 | 1102 | |||
GenBank mRNA | BC037450.1 | - | 6 | 707 | ||
D50060.1 | - | 8 | 1025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
