siRNA Details
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291184.1 | NP_001278113.1 | 10 | 1408 | ||
NM_011048.1 | NP_035178.1 | 10 | 1408 | |||
GenBank mRNA | BC037450.1 | - | 8 | 1013 | ||
D50060.1 | - | 10 | 1331 |

More Information
Other Species Targeted

Gene Ontology Categories:
