siRNA Details
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008795.2 | NP_032821.1 | 4, 5 | 642 | ||
GenBank mRNA | AK004998.1 | - | 4, 5 | 586 | ||
AK171569.1 | - | 4, 5 | 423 | |||
AK198726.1 | - | 1, 2 | 101 | |||
BC119617.1 | - | 5, 6 | 518 | 40061728 | ||
X69026.1 | - | 4, 5 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
